Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Boc-3,4-dihydro-2H-pyridine
N-Boc-3,4-dihydro-2H-pyridine
N-Boc-3,4-dihydro-2H-pyridine
N-Boc-3,4-dihydro-2H-pyridine
APOSOR47889-1G
:
Apollo Scientific
:
N-Boc-3,4-dihydro-2H-pyridine
APOSOR47889-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
183.247 g/mol
:
Ambient
:
2810
:
6.1,III
:
131667-57-7
:
C₁₀H₁₇NO₂
:
MFCD00798175