Order Entry
Export
ContactUsLinkComponent
4-Bromopyridine-2,6-dicarboxamide
Irritant
4-Bromopyridine-2,6-dicarboxamide
Catalog # APOSOR4731-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromopyridine-2,6-dicarboxamide
Catalog # APOSOR4731-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :244.045 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:668992-72-1
Formula:C₇H₆BrN₃O₂
MDL Number:MFCD04113814