Order Entry
Export
ContactUsLinkComponent
 
(S)-1,2-Dimethylpiperazine 95%
Catalog # APOSOR471705-1G
Supplier:  Apollo Scientific
CAS Number:  
(S)-1,2-Dimethylpiperazine 95%
Catalog # APOSOR471705-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :114.189 g/mol
Storage Temperature:Ambient
CAS Number:485841-52-9
Formula:C₆H₁₄N₂
MDL Number:MFCD11858472