Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Chloro-4-methoxyindoline-2,3-dione
7-Chloro-4-methoxyindoline-2,3-dione
7-Chloro-4-methoxyindoline-2,3-dione
7-Chloro-4-methoxyindoline-2,3-dione
Supplier:
Apollo Scientific
Certificates
Expand image
7-Chloro-4-methoxyindoline-2,3-dione
Supplier:
Apollo Scientific
About this item
CAS Number:
15345-55-8
:
Formula:
C₉H₆ClNO₃
MDL Number:
MFCD00835536
:
211.602 g/mol
Storage Temperature:
Ambient