Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Dimethyl-N-(4-methyl-2-pyridyl)propionamide
2,2-Dimethyl-N-(4-methyl-2-pyridyl)propionamide
2,2-Dimethyl-N-(4-methyl-2-pyridyl)propionamide
2,2-Dimethyl-N-(4-methyl-2-pyridyl)propionamide
Supplier:
Apollo Scientific
Certificates
Expand image
2,2-Dimethyl-N-(4-methyl-2-pyridyl)propionamide
Supplier:
Apollo Scientific
About this item
CAS Number:
86847-77-0
:
Formula:
C₁₁H₁₆N₂O
MDL Number:
MFCD00996305
:
192.258 g/mol
Storage Temperature:
Ambient