
Methyl 12-Bromododecanoate
Catalog # APOSOR471516-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:293.24 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:26825-95-6
Formula:C₁₃H₂₅BrO₂
MDL Number:MFCD01838179