Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(4-Bromophenyl)oxazolo[4,5-c]pyridine
2-(4-Bromophenyl)oxazolo[4,5-c]pyridine
2-(4-Bromophenyl)oxazolo[4,5-c]pyridine
2-(4-Bromophenyl)oxazolo[4,5-c]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
2-(4-Bromophenyl)oxazolo[4,5-c]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
934331-04-1
:
Formula:
C₁₂H₇BrN₂O
MDL Number:
MFCD22573880
:
275.101 g/mol
Storage Temperature:
Ambient