Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
[6-(3,5-Dichlorophenyl)-3-pyridyl]methanol
[6-(3,5-Dichlorophenyl)-3-pyridyl]methanol
[6-(3,5-Dichlorophenyl)-3-pyridyl]methanol
[6-(3,5-Dichlorophenyl)-3-pyridyl]methanol
Supplier:
Apollo Scientific
Certificates
Expand image
[6-(3,5-Dichlorophenyl)-3-pyridyl]methanol
Supplier:
Apollo Scientific
About this item
CAS Number:
887974-84-7
:
Formula:
C₁₂H₉Cl₂NO
MDL Number:
MFCD06410460
:
254.112 g/mol
Storage Temperature:
Ambient