2-(Hydroxymethyl)-1H-imidazo[4,5-b]phenazine
Catalog # APOSOR471355-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:250.255 g/mol
Storage Temperature:Ambient
CAS Number:2006276-99-7
Formula:C₁₄H₁₀N₄O
MDL Number:MFCD28977368