Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dimethylimidazole
4,5-Dimethylimidazole
4,5-Dimethylimidazole
:
Apollo Scientific
4,5-Dimethylimidazole
:
Apollo Scientific
:
2302-39-8
:
:
C₅H₈N₂
:
MFCD01646351
:
96.13 g/mol
:
GHS07
:
Ambient