Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,6-Dimethylpyrimidin-4(3H)-one
5,6-Dimethylpyrimidin-4(3H)-one
5,6-Dimethylpyrimidin-4(3H)-one
5,6-Dimethylpyrimidin-4(3H)-one
Supplier:
Apollo Scientific
Certificates
Expand image
5,6-Dimethylpyrimidin-4(3H)-one
Supplier:
Apollo Scientific
About this item
CAS Number:
34916-78-4
:
MFCD00234217
Formula:
C₆H₈N₂O
MDL Number:
MFCD14605439
:
124.141 g/mol
Storage Temperature:
Ambient