2-(Aminomethyl)-3,5-dimethylpyrrole
Catalog # APOSOR471267-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:124.184 g/mol
Storage Temperature:Ambient
CAS Number:1503251-51-1
Formula:C₇H₁₂N₂
MDL Number:MFCD28398135