N-(3,5-Dimethoxybenzyl)cyclopropanamine
Catalog # APOSOR471171-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:207.269 g/mol
Storage Temperature:Ambient
CAS Number:625435-20-3
Formula:C₁₂H₁₇NO₂
MDL Number:MFCD11131159