2,2'-Dinitrobenzidine
Catalog # APOSOR471107-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:274.232 g/mol
Storage Temperature:Ambient
CAS Number:5855-71-0
Formula:C₁₂H₁₀N₄O₄
MDL Number:MFCD02752221