2,6-Dihydroxyhexanenitrile
Catalog # APOSOR471070-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:129.157 g/mol
Storage Temperature:Ambient
CAS Number:17264-36-7
Formula:C₆H₁₁NO₂
MDL Number:MFCD19300782