Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2,4-Trimethyl-1,2-dihydroquinoline
2,2,4-Trimethyl-1,2-dihydroquinoline
2,2,4-Trimethyl-1,2-dihydroquinoline
2,2,4-Trimethyl-1,2-dihydroquinoline
Supplier:
Apollo Scientific
Certificates
Expand image
2,2,4-Trimethyl-1,2-dihydroquinoline
Supplier:
Apollo Scientific
About this item
CAS Number:
147-47-7
:
Formula:
C₁₂H₁₅N
MDL Number:
MFCD00044248
:
173.254 g/mol
Storage Temperature:
Ambient