Methyl 3-hydroxy-4,4-dimethylpentanoate
Catalog # APOSOR471046-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:160.211 g/mol
Storage Temperature:Ambient
CAS Number:150943-32-1
Formula:C₈H₁₆O₃
MDL Number:MFCD28101621