(S)-4-Benzyl-2-(hydroxymethyl)morpholine
Catalog # APOSOR471039-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:207.269 g/mol
Storage Temperature:Ambient
CAS Number:132073-82-6
Formula:C₁₂H₁₇NO₂
MDL Number:MFCD10698663