3,6-Dichloro-4-iodopyridazine
Catalog # APOSOR470888-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:274.875 g/mol
Storage Temperature:Ambient
CAS Number:130825-13-7
Formula:C₄HCl₂IN₂
MDL Number:MFCD18249972