Order Entry
Export
ContactUsLinkComponent
cis-3,7-Diazabicyclo[3.3.0]octane
cis-3,7-Diazabicyclo[3.3.0]octane
Catalog # APOSOR470881-1G
Supplier:  Apollo Scientific
CAS Number:  
cis-3,7-Diazabicyclo[3.3.0]octane
Catalog # APOSOR470881-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :112.173 g/mol
Storage Temperature:Ambient
CAS Number:19885-60-0
Formula:C₆H₁₂N₂
MDL Number:MFCD27935365