Order Entry
Export
ContactUsLinkComponent
3-Bromo-6-(methoxymethoxy)-o-xylene
3-Bromo-6-(methoxymethoxy)-o-xylene
Catalog # APOSOR470856-5G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-6-(methoxymethoxy)-o-xylene
Catalog # APOSOR470856-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :245.113 g/mol
Storage Temperature:Ambient
CAS Number:1301147-34-1
Formula:C₁₀H₁₃BrO₂
MDL Number:MFCD27935368