Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Methoxymethoxy)-2,3-dimethylbenzoic acid
4-(Methoxymethoxy)-2,3-dimethylbenzoic acid
4-(Methoxymethoxy)-2,3-dimethylbenzoic acid
4-(Methoxymethoxy)-2,3-dimethylbenzoic acid
Supplier:
Apollo Scientific
Certificates
Expand image
4-(Methoxymethoxy)-2,3-dimethylbenzoic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
1333231-02-9
:
Formula:
C₁₁H₁₄O₄
MDL Number:
MFCD24501863
:
210.226 g/mol
Storage Temperature:
Ambient