2-Cbz-2,7-diazaspiro[4.5]decane
Catalog # APOSOR470850-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:274.358 g/mol
Storage Temperature:Ambient
CAS Number:1086394-72-0
Formula:C₁₆H₂₂N₂O₂
MDL Number:MFCD11223504