2-Cbz-2,8-diazaspiro[5.5]undecane
Catalog # APOSOR470847-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:288.385 g/mol
Storage Temperature:Ambient
CAS Number:1086394-63-9
Formula:C₁₇H₂₄N₂O₂
MDL Number:MFCD11223514