Order Entry
Export
ContactUsLinkComponent
8-Cbz-1,8-diazaspiro[5.5]undecane
8-Cbz-1,8-diazaspiro[5.5]undecane
Catalog # APOSOR470845-1G
Supplier:  Apollo Scientific
CAS Number:  
8-Cbz-1,8-diazaspiro[5.5]undecane
Catalog # APOSOR470845-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :288.385 g/mol
Storage Temperature:Ambient
CAS Number:1086394-89-9
Formula:C₁₇H₂₄N₂O₂
MDL Number:MFCD11223536