Order Entry
Export
ContactUsLinkComponent
1'-Boc-spiro[indene-1,4'-piperidine]
1'-Boc-spiro[indene-1,4'-piperidine]
Catalog # APOSOR470774-5G
Supplier:  Apollo Scientific
CAS Number:  
1'-Boc-spiro[indene-1,4'-piperidine]
Catalog # APOSOR470774-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :285.381 g/mol
Storage Temperature:Ambient
CAS Number:137419-24-0
Formula:C₁₈H₂₃NO₂
MDL Number:MFCD03160565