Order Entry
Export
ContactUsLinkComponent
3,5-Dibromo-4-chlorophenol
3,5-Dibromo-4-chlorophenol
Catalog # APOSOR470759-5G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Dibromo-4-chlorophenol
Catalog # APOSOR470759-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :286.348 g/mol
Storage Temperature:Ambient
CAS Number:74104-19-1
Formula:C₆H₃Br₂ClO
MDL Number:MFCD18914596