Order Entry
Export
ContactUsLinkComponent
2-Phenylimidazo[1,2-a]pyridine-7-carboxylic acid
2-Phenylimidazo[1,2-a]pyridine-7-carboxylic acid
Catalog # APOSOR470753-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Phenylimidazo[1,2-a]pyridine-7-carboxylic acid
Catalog # APOSOR470753-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :238.241 g/mol
Storage Temperature:Ambient
CAS Number:959-40-0
Formula:C₁₄H₁₀N₂O₂
MDL Number:MFCD08164306