Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2,5-Diaminobenzoate
Methyl 2,5-Diaminobenzoate
Methyl 2,5-Diaminobenzoate
Methyl 2,5-Diaminobenzoate
APOSOR470750-1G
:
Apollo Scientific
:
Methyl 2,5-Diaminobenzoate
APOSOR470750-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
166.177 g/mol
:
Ambient
:
49592-84-9
:
C₈H₁₀N₂O₂
:
MFCD12913485