Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,7-Dimethyl-1,4-diazepane
1,7-Dimethyl-1,4-diazepane
1,7-Dimethyl-1,4-diazepane
1,7-Dimethyl-1,4-diazepane
APOSOR470722-1G
:
Apollo Scientific
:
1,7-Dimethyl-1,4-diazepane
APOSOR470722-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
128.215 g/mol
:
Ambient
:
67744-48-3
:
C₇H₁₆N₂
:
MFCD19221307