Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-Methyl-3-oxo-3-phenylpropanoate
Methyl 2-Methyl-3-oxo-3-phenylpropanoate
Methyl 2-Methyl-3-oxo-3-phenylpropanoate
APOSOR470721-5G
:
Apollo Scientific
:
Methyl 2-Methyl-3-oxo-3-phenylpropanoate
APOSOR470721-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
192.211 g/mol
:
Ambient
:
29540-54-3
:
C₁₁H₁₂O₃
:
MFCD00956709