1H-Imidazo[4,5-b]pyrazine
Catalog # APOSOR470676-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:120.112 g/mol
Storage Temperature:Ambient
CAS Number:273-94-9
Formula:C₅H₄N₄
MDL Number:MFCD08669465