Ethyl 2-(4,6-Dichloro-5-pyrimidyl)acetate
Catalog # APOSOR470670-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:235.067 g/mol
Storage Temperature:Ambient
CAS Number:1095822-21-1
Formula:C₈H₈Cl₂N₂O₂
MDL Number:MFCD13195351