2-Chloro-2'-hydroxy-3'-methylacetophenone
Catalog # APOSOR470663-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.62 g/mol
Storage Temperature:Ambient
CAS Number:75717-51-0
Formula:C₉H₉ClO₂
MDL Number:MFCD13172653