Ethyl 4,4-Dimethyl-2-oxopyrrolidine-3-carboxylate
Catalog # APOSOR470656-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:185.22 g/mol
Storage Temperature:Ambient
CAS Number:90609-07-7
Formula:C₉H₁₅NO₃
MDL Number:MFCD12828110