Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Nitro-1,3-dihydroisobenzofuran
4-Nitro-1,3-dihydroisobenzofuran 95%
4-Nitro-1,3-dihydroisobenzofuran 95%
4-Nitro-1,3-dihydroisobenzofuran 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Nitro-1,3-dihydroisobenzofuran 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
98475-08-2
:
Formula:
C₈H₇NO₃
MDL Number:
MFCD16659157
:
165.146 g/mol
Storage Temperature:
Ambient