Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 3-(3-Pyridyl)propiolate
Ethyl 3-(3-Pyridyl)propiolate
Ethyl 3-(3-Pyridyl)propiolate
APOSOR470614-5G
:
Apollo Scientific
:
Ethyl 3-(3-Pyridyl)propiolate
APOSOR470614-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
175.184 g/mol
:
Ambient
:
59608-03-6
:
C₁₀H₉NO₂
:
MFCD01073808