4-(Trimethylsilyl)phenyl Pivalate
Catalog # APOSOR470570-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:250.409 g/mol
Storage Temperature:Ambient
CAS Number:1418117-88-0
Formula:C₁₄H₂₂O₂Si
MDL Number:MFCD22682787