Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-(Trimethylsilyl)benzoate
Methyl 4-(Trimethylsilyl)benzoate
Methyl 4-(Trimethylsilyl)benzoate
Methyl 4-(Trimethylsilyl)benzoate
APOSOR470568-5G
:
Apollo Scientific
:
Methyl 4-(Trimethylsilyl)benzoate
APOSOR470568-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
208.329 g/mol
:
Ambient
:
22515-30-6
:
C₁₁H₁₆O₂Si
:
MFCD22682799