Methyl 1-(3,4-Dichlorophenyl)-4-oxocyclohexanecarboxylate
Catalog # APOSOR470539-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:301.165 g/mol
Storage Temperature:Ambient
CAS Number:1384264-84-9
Formula:C₁₄H₁₄Cl₂O₃
MDL Number:MFCD22423147