Order Entry
Export
ContactUsLinkComponent
Methyl 1-(3,4-Dimethoxyphenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(3,4-Dimethoxyphenyl)-4-oxocyclohexanecarboxylate
Catalog # APOSOR470516-5G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 1-(3,4-Dimethoxyphenyl)-4-oxocyclohexanecarboxylate
Catalog # APOSOR470516-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :292.327 g/mol
Storage Temperature:Ambient
CAS Number:1385694-65-4
Formula:C₁₆H₂₀O₅
MDL Number:MFCD22423114