α-Acetoxy-(3,4-dimethoxyphenyl)acetonitrile
Catalog # APOSOR470508-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:235.236 g/mol
Storage Temperature:Ambient
CAS Number:13782-40-6
Formula:C₁₂H₁₃NO₄
MDL Number:MFCD20128188