Order Entry
Export
ContactUsLinkComponent
(1R,6S)-3-Methyl-3,8-diazabicyclo[4.2.0]octane
(1R,6S)-3-Methyl-3,8-diazabicyclo[4.2.0]octane
Catalog # APOSOR470483-250MG
Supplier:  Apollo Scientific
CAS Number:  
(1R,6S)-3-Methyl-3,8-diazabicyclo[4.2.0]octane
Catalog # APOSOR470483-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :126.199 g/mol
Storage Temperature:Ambient
CAS Number:1434126-84-7
Formula:C₇H₁₄N₂
MDL Number:MFCD22205854