Ethyl 2-(2-Benzoxazolylamino)-6-oxo-1,6-dihydropyrimidine-5-carboxylate
Catalog # APOSOR470448-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:300.269 g/mol
Storage Temperature:Ambient
CAS Number:670220-69-6
Formula:C₁₄H₁₂N₄O₄
MDL Number:MFCD22205841