
3,4-Dihydro-4,4-dimethyl-1,3,5-triazino[1,2-a]benzimidazol-2-amine
Catalog # APOSOR470443-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:215.254 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:325738-83-8
Formula:C₁₁H₁₃N₅
MDL Number:MFCD02323006