2-Amino-2-(3,4,5-trimethoxyphenyl)acetic acid hydrochloride
Catalog # APOSOR470406-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:277.701 g/mol
Storage Temperature:Ambient
CAS Number:86053-95-4
Formula:C₁₁H₁₅NO₅·ClH
MDL Number:MFCD06410544