1-(3,4-Dimethoxyphenyl)-4-oxocyclohexanecarboxylic acid
Catalog # APOSOR470394-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:278.3 g/mol
Storage Temperature:Ambient
CAS Number:887978-64-5
Formula:C₁₅H₁₈O₅
MDL Number:MFCD06410575