4-Acetamido-3,5-dimethylphenol
Catalog # APOSOR470383-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:179.216 g/mol
Storage Temperature:Ambient
CAS Number:6337-56-0
Formula:C₁₀H₁₃NO₂
MDL Number:MFCD00748645