N-(4-Bromobenzyl)-2,2-dimethoxyethylamine
Catalog # APOSOR470380-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:274.154 g/mol
Storage Temperature:Ambient
CAS Number:1036378-89-8
Formula:C₁₁H₁₆BrNO₂
MDL Number:MFCD12148381