2-Amino-2-(3,5-dichlorophenyl)acetic acid hydrochloride
Catalog # APOSOR470326-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:256.514 g/mol
Storage Temperature:Ambient
CAS Number:1137014-87-9
Formula:C₈H₇Cl₂NO₂·ClH
MDL Number:MFCD06202321